missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthylacetic Acid, TRC

Product Code. 30468552
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30468552 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30468552 Supplier Toronto Research Chemicals Supplier No. TRCN3684905G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Naphthyl acetic acid
CAS 581-96-4
Molecular Formula C12 H10 O2
Quantity 5 g
Synonym 2-(2-Naphthyl)acetic acid, 2-NAA, 2-Naphthalen-2-ylacetic acid, 2-Naphthylacetic acid, Betoxan, NSC 301, Naphthalen-2-ylacetic acid, β-Naphthaleneacetate, β-Naphthaleneacetic acid, β-Naphthylacetic acid
InChI Formula InChI=1S/C12H10O2/c13-12(14)8-9-5-6-10-3-1-2-4-11(10)7-9/h1-7H,8H2,(H,13,14)
SMILES OC(=O)Cc1ccc2ccccc2c1
IUPAC Name 2-naphthalen-2-ylacetic acid
Molecular Weight (g/mol) 186.21
Formula Weight 186.0681
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.