missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthyl trifluoromethanesulfonate, 97%, Thermo Scientific™

Product Code. 15431648
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15431648

Brand: Thermo Scientific Alfa Aesar H55417.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3857-83-8
Molecular Formula C11H7F3O3S
Molecular Weight (g/mol) 276.229
MDL Number MFCD00192341
InChI Key MDWRQYBWVTXIIJ-UHFFFAOYSA-N
Synonym 2-naphthyl trifluoromethanesulfonate, 2-naphthyl triflate, 2-naphthyltrifluoromethanesulfonate, 2-naphthyltriflate, acmc-1aigh, trifluoromethanesulfonic acid 2-naphthyl ester, methanesulfonic acid, 1,1,1-trifluoro-, 2-naphthalenyl ester, methanesulfonic acid,1,1,1-trifluoro-, 2-naphthalenyl ester
PubChem CID 4546341
IUPAC Name naphthalen-2-yl trifluoromethanesulfonate
SMILES C1=CC=C2C=C(C=CC2=C1)OS(=O)(=O)C(F)(F)F

Specifications

Melting Point 30°C to 32°C
Color Orange to White
Flash Point 113°C (235°F)
Quantity 1 g
UN Number UN3261
Formula Weight 276.23
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Naphthyl trifluoromethanesulfonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.