missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthyl phenyl ketone, 98%, Thermo Scientific™

Catalog Number p-171197
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
644-13-3
C17H12O
232.28
MFCD00004106
SJNXJRVDSTZUFB-UHFFFAOYSA-N
2-benzoylnaphthalene, 2-naphthyl phenyl ketone, 2-benzonaphthone, naphthalen-2-yl phenyl methanone, methanone, 2-naphthalenylphenyl, 2'-benzonaphthone, ketone, 2-naphthyl phenyl, 2-naphthylphenylketone, beta-benzoylnaphthalene, 2-naphthyl phenyl methanone
69516
naphthalen-2-yl(phenyl)methanone
O=C(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1
This item is not returnable. View return policy
644-13-3
C17H12O
232.28
MFCD00004106
SJNXJRVDSTZUFB-UHFFFAOYSA-N
2-benzoylnaphthalene, 2-naphthyl phenyl ketone, 2-benzonaphthone, naphthalen-2-yl phenyl methanone, methanone, 2-naphthalenylphenyl, 2'-benzonaphthone, ketone, 2-naphthyl phenyl, 2-naphthylphenylketone, beta-benzoylnaphthalene, 2-naphthyl phenyl methanone
69516
naphthalen-2-yl(phenyl)methanone
O=C(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1
CAS Min % 97.5
CAS Max % 100.0
Melting Point 76°C to 82°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 232.28
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Naphthyl phenyl ketone, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'