missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthyl isocyanate, 97%, Thermo Scientific™

Product Code. 15467957
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15467957

Brand: Thermo Scientific Alfa Aesar H51076.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Naphthyl isocyanate is used as pharmaceutical intermediate.

Solubility
It hydrolyzes in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, alcohols and amines.
TRUSTED_SUSTAINABILITY

Specifications

CAS 2243-54-1
Melting Point 53°C to 56°C
Boiling Point 83°C (0.2 mmHg)
Odor Pungent
Molecular Formula C11H7NO
MDL Number MFCD00014319
Quantity 1 g
UN Number UN2811
Sensitivity Moisture sensitive
Synonym 2-naphthyl isocyanate, 2-naphthylisocyanate, naphthalene, 2-isocyanato, beta-naphthylisocyanate, unii-9ze6124voi, naphthalene,2-isocyanato, naphthalen-2-isocyanate, 2-naphthalenyl isocyanate, 2-isocyanato-naphthalene, acmc-1cr5l
Solubility Information It hydrolyzes in water.
InChI Key XIXJQNFTNSQTBT-UHFFFAOYSA-N
SMILES C1=CC=C2C=C(C=CC2=C1)N=C=O
IUPAC Name 2-isocyanatonaphthalene
Molecular Weight (g/mol) 169.183
PubChem CID 16718
Formula Weight 169.18
Percent Purity 97%
Chemical Name or Material 2-Naphthyl isocyanate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.