missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthonitrile, 97%

Product Code. 10246051 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10246051 10 g Glass bottle 10g
10397461 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10246051 Supplier Thermo Scientific Acros Supplier No. 346310100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 613-46-7 | C11H7N | 153.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 613-46-7
Molecular Formula C11H7N
Molecular Weight (g/mol) 153.18
MDL Number MFCD00016807
InChI Key AZKDTTQQTKDXLH-UHFFFAOYSA-N
Synonym 2-naphthonitrile, 2-cyanonaphthalene, 2-naphthalenecarbonitrile, 2-cyanonaphthylene, 2-naphthalenenitrile, beta-cyanonaphthalene, beta-naphthonitrile, .beta.-naphthonitrile, naphthalene-2-carbonitrile, 2-naphthylcyanide
PubChem CID 11944
IUPAC Name naphthalene-2-carbonitrile
SMILES C1=CC=C2C=C(C=CC2=C1)C#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 63°C to 68°C
Color White to Beige
Boiling Point 156°C to 158°C (12.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 10 g
Formula Weight 153.18
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Naphthonitrile, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Naphthonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.