missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthol, Extra Pure, SLR, Fisher Chemical

Product Code. 10528630 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10528630 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10528630 Supplier Fisher Chemical Supplier No. N/0400/53

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 135-19-3
Melting Point 120°C
Color Beige
pH 5.7
Boiling Point 285°C
Packaging Plastic powder jar
Molecular Formula C10H8O
MDL Number 4067
Quantity 500 g
Synonym 2-naphthol, 2-naphthalenol, beta-naphthol, 2-hydroxynaphthalene, betanaphthol, isonaphthol, developer bn, naphthol b, azogen developer a, beta-napthol
InChI Key JWAZRIHNYRIHIV-UHFFFAOYSA-N
SMILES C1=CC=C2C=C(C=CC2=C1)O
IUPAC Name naphthalen-2-ol
Molecular Weight (g/mol) 144.173
PubChem CID 8663
ChEBI CHEBI:10432
Formula Weight 144.17g/mol
Physical Form Solid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Naphthol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.