missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthol, 99+%

Product Code. 10276753 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
100g
10kg
1kg
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10276753

Brand: Thermo Scientific Acros 180820010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 135-19-3 | C10H8O | 144.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-19-3
Molecular Formula C10H8O
Molecular Weight (g/mol) 144.17
MDL Number MFCD00004067
InChI Key JWAZRIHNYRIHIV-UHFFFAOYSA-N
Synonym 2-naphthol, 2-naphthalenol, beta-naphthol, 2-hydroxynaphthalene, betanaphthol, isonaphthol, developer bn, naphthol b, azogen developer a, beta-napthol
PubChem CID 8663
ChEBI CHEBI:10432
IUPAC Name naphthalen-2-ol
SMILES C1=CC=C2C=C(C=CC2=C1)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 120.0°C to 124.0°C
Color White
Boiling Point 285.0°C to 286.0°C
Flash Point 153°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C10H7OH
Quantity 1 kg
Beilstein 06,627
Merck Index 15,6469
Solubility Information Solubility in water: 1g/L (20°C). Other solubilities: 1 g/80mL boiling water, 1 g/0.8mL alcohol, 1 g/17mL chloroform, 1 g/1.3mL ether, soluble in ethanol, ether, chloroform, glycerol
Formula Weight 144.17
Percent Purity 99+%
Physical Form Crystals, Powder or Granules
Chemical Name or Material 2-Naphthol, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Naphthol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.