missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthol, 98+%

Product Code. 11491948
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491948 1000 g 1000g
11401958 250 g 250g
11411958 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491948 Supplier Thermo Scientific Alfa Aesar Supplier No. A14564.0B
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 135-19-3 | C10H8O | 144.173 g/mol

2-Naphthol can be used as a fluorescent indicator. It can be also used in the production of dyes and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Naphthol can be used as a fluorescent indicator. It can be also used in the production of dyes and in organic synthesis.

Solubility
Partly miscible in water (0.76 g/L).

Notes
It is sensitive to light. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-19-3
Molecular Formula C10H8O
Molecular Weight (g/mol) 144.173
MDL Number MFCD00004067
InChI Key JWAZRIHNYRIHIV-UHFFFAOYSA-N
Synonym 2-naphthol, 2-naphthalenol, beta-naphthol, 2-hydroxynaphthalene, betanaphthol, isonaphthol, developer bn, naphthol b, azogen developer a, beta-napthol
PubChem CID 8663
ChEBI CHEBI:10432
IUPAC Name naphthalen-2-ol
SMILES C1=CC=C2C=C(C=CC2=C1)O

Specifications

Melting Point 120°C to 124°C
Density 1.28
pH 5.77
Boiling Point 285°C to 286°C
Flash Point 153°C (307°F)
Odor Phenol-like
Quantity 1000 g
UN Number UN3077
Beilstein 742134
Sensitivity Light sensitive
Merck Index 14,6384
Solubility Information Partly miscible in water (0.76g/L).
Formula Weight 144.17
Percent Purity ≥98%
Chemical Name or Material 2-Naphthol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.