missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthoic acid, 98+%

Product Code. 11381827
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11381827 25 g 25g
11371827 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11381827 Supplier Thermo Scientific Alfa Aesar Supplier No. L05102.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 93-09-4 | C11H8O2 | 172.183 g/mol

2-Naphthoic acid is an unprotected acid undergoes lithiation, e.g. with s-BuLi, by stereospecific 1,4-addition, providing a facile route to 1,2,2-trisubstituted 1,2-dihydronaphthalenes. It is also used as an intermediate in organic synthesis. The fluorescence spectra and electronic absorption of 2-naphthoic acid was studied.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Naphthoic acid is an unprotected acid undergoes lithiation, e.g. with s-BuLi, by stereospecific 1,4-addition, providing a facile route to 1,2,2-trisubstituted 1,2-dihydronaphthalenes. It is also used as an intermediate in organic synthesis. The fluorescence spectra and electronic absorption of 2-naphthoic acid was studied.

Solubility
Soluble in alcohol, diethyl ether, methanol, and ethyl acetate (20 mg/ml). Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-09-4
Molecular Formula C11H8O2
Molecular Weight (g/mol) 172.183
MDL Number MFCD00004101
InChI Key UOBYKYZJUGYBDK-UHFFFAOYSA-N
Synonym 2-naphthoic acid, 2-naphthalenecarboxylic acid, isonaphthoic acid, 2-carboxynaphthalene, 2-maythic acid, beta-naphthoic acid, ne-2-carboxylic acid, unii-qlg01v0w2l, naphthalene-beta-carboxylic acid, .beta.-naphthoic acid
PubChem CID 7123
ChEBI CHEBI:36106
IUPAC Name naphthalene-2-carboxylic acid
SMILES C1=CC=C2C=C(C=CC2=C1)C(=O)O

Specifications

Melting Point 182°C to 186°C
Density 1.08
Flash Point 205°C (401°F)
Quantity 25 g
Beilstein 972039
Merck Index 14,6382
Solubility Information Soluble in alcohol,diethyl ether,methanol,and ethyl acetate (20mg/ml). Insoluble in water.
Formula Weight 172.18
Percent Purity ≥98%
Chemical Name or Material 2-Naphthoic acid
compliance-icons
Product Identifier
  • 2-Naphthoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-Naphthalenecarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.