missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthalenemethanol, 98%

Product Code. 10282842 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10282842 5 g Glass bottle 5g
10724743 1 g Glass bottle 1g
10646564 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10282842 Supplier Thermo Scientific Acros Supplier No. 351150050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1592-38-7 | C11H10O | 158.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1592-38-7
Molecular Formula C11H10O
Molecular Weight (g/mol) 158.2
MDL Number MFCD00004124
InChI Key MFGWMAAZYZSWMY-UHFFFAOYSA-N
Synonym 2-naphthalenemethanol, 2-naphthylmethanol, 2-hydroxymethylnaphthalene, 2-naphthalene methanol, 2-naphthyl methanol, naphthalen-2-yl-methanol, naphthalen-2-yl methanol, 2-naphthylmethan-1-ol, 2-naphthylcarbinol, 2na
PubChem CID 74128
ChEBI CHEBI:27615
IUPAC Name naphthalen-2-ylmethanol
SMILES C1=CC=C2C=C(C=CC2=C1)CO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 79°C to 83°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C10H7CH2OH
Quantity 5 g
Formula Weight 158.2
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Naphthalenemethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.