missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthaldehyde, 98%

Product Code. 11486237
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486237 100 g 100g
11466237 5 g 5g
11476237 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11486237 Supplier Thermo Scientific Alfa Aesar Supplier No. B25157.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 66-99-9 | C11H8O | 156.184 g/mol

2-Naphthaldehyde aids in stereo-selective formation of bicyclo 2-siloxy-2-alkoxyoxetanes. It is also used as catalytic agent and petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Naphthaldehyde aids in stereo-selective formation of bicyclo 2-siloxy-2-alkoxyoxetanes. It is also used as catalytic agent and petrochemical additive.

Solubility
Soluble in hot water.

Notes
Store at -20°C. Air Sensitive. Store away from string bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-99-9
Molecular Formula C11H8O
Molecular Weight (g/mol) 156.184
MDL Number MFCD00004094
InChI Key PJKVFARRVXDXAD-UHFFFAOYSA-N
Synonym 2-naphthaldehyde, 2-naphthalenecarboxaldehyde, 2-formylnaphthalene, beta-naphthaldehyde, beta-formylnaphthalene, beta-naphthylaldehyde, beta-naphthylcarboxaldehyde, .beta.-naphthaldehyde, .beta.-naphthylaldehyde, .beta.-formylnaphthalene
PubChem CID 6201
ChEBI CHEBI:52368
IUPAC Name naphthalene-2-carbaldehyde
SMILES C1=CC=C2C=C(C=CC2=C1)C=O

Specifications

Melting Point 59°C to 62°C
Boiling Point 160°C (19 mmHg)
Quantity 100 g
Beilstein 507750
Sensitivity Air Sensitive
Solubility Information Soluble in hot water.
Formula Weight 156.18
Percent Purity 98%
Chemical Name or Material 2-Naphthaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.