missing translation for 'onlineSavingsMsg'
Learn More

2-(n-Propylthio)nicotinic acid, 97%, Thermo Scientific™

Product Code. 11407576
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
missing translation for 'unitSize'
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11407576

missing translation for 'mfr': Thermo Scientific Alfa Aesar A14623.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175135-22-5
Molecular Formula C9H10NO2S
Molecular Weight (g/mol) 196.24
MDL Number MFCD00051682
InChI Key CSMDLVRDBYKTAH-UHFFFAOYSA-M
Synonym 2-propylthio nicotinic acid, 2-n-propylthio nicotinic acid, 2-propylsulfanyl pyridine-3-carboxylic acid, 3-pyridinecarboxylicacid, 2-propylthio, 2-propylthiopyridine-3-carboxylic acid, maybridge1_001579, 2-prop-1-ylsulphanyl pyridine-3-carboxylic acid, 2-propylsulfanyl-nicotinic acid
PubChem CID 2774526
IUPAC Name 2-propylsulfanylpyridine-3-carboxylic acid
SMILES CCCSC1=NC=CC=C1C([O-])=O

Specifications

Melting Point 159°C to 163°C
Quantity 5 g
Formula Weight 197.26
Percent Purity 97%
Chemical Name or Material 2-(n-Propylthio)nicotinic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.