missing translation for 'onlineSavingsMsg'
Learn More

2-n-Propylphenol, 97%, Thermo Scientific™

Catalog Number p-7081211
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy
644-35-9
C9H12O
136.194
MFCD00002251
LCHYEKKJCUJAKN-UHFFFAOYSA-N
2-n-propylphenol, o-propylphenol, phenol, 2-propyl, phenol, o-propyl, 1-hydroxy-2-n-propylbenzene, 1-2-hydroxyphenyl propane, propylphenol, 2-propyl-phenol, o-n-propylphenol, unii-333r6f6t3n
12570
2-propylphenol
CCCC1=CC=CC=C1O
This item is not returnable. View return policy
644-35-9
C9H12O
136.194
MFCD00002251
LCHYEKKJCUJAKN-UHFFFAOYSA-N
2-n-propylphenol, o-propylphenol, phenol, 2-propyl, phenol, o-propyl, 1-hydroxy-2-n-propylbenzene, 1-2-hydroxyphenyl propane, propylphenol, 2-propyl-phenol, o-n-propylphenol, unii-333r6f6t3n
12570
2-propylphenol
CCCC1=CC=CC=C1O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 0.989
Boiling Point 214°C
Flash Point 93°C (199°F)
Odor Phenol-like
Refractive Index 1.528
Quantity 25 g
UN Number UN3145
Beilstein 1363932
Formula Weight 136.19
Percent Purity 97%
Chemical Name or Material 2-n-Propylphenol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'