missing translation for 'onlineSavingsMsg'
Learn More

2-n-Propyl-1-pentanol, 97%

Product Code. 15494338
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15494338 1 g 1g
15404348 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15494338 Supplier Thermo Scientific Alfa Aesar Supplier No. H57954.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 58175-57-8 | C8H18O | 130.231 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58175-57-8
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.231
MDL Number MFCD00010260
InChI Key LASHFHLFDRTERB-UHFFFAOYSA-N
Synonym 2-propyl-1-pentanol, 1-pentanol, 2-propyl, 2-propylpentanol, 2-n-propyl-1-pentanol, acmc-1azt3, 2-propyl-1-hydroxypentane
PubChem CID 123543
IUPAC Name 2-propylpentan-1-ol
SMILES CCCC(CCC)CO

Specifications

Color Colorless to Yellow
Density 0.83 g/mL
Boiling Point 80°C to 82°C (12 mmHg)
Flash Point 72°C (162°F)
Linear Formula (CH3CH2CH2)2CHCH2OH
Refractive Index 1.43
Quantity 1 g
Formula Weight 130.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-n-Propyl-1-pentanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.