missing translation for 'onlineSavingsMsg'
Learn More

2-n-Propoxybenzoic acid, 98+%, Thermo Scientific™

Catalog Number 11450428
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
2100-31-4
C10H12O3
180.203
MFCD01075687
OXOWWPXTTOCKKU-UHFFFAOYSA-N
2-n-propoxybenzoic acid, o-propoxybenzoic acid, benzoic acid, 2-propoxy, 2-propoxy benzoic acid, salicylic acid propyl ether, akos bbb/270, rarechem al bo 0876, propyloxybenzoic acid, 2-propoxybenzoic acid, o-propoxy benzoic acid
539215
2-propoxybenzoic acid
CCCOC1=CC=CC=C1C(=O)O
This item is not returnable. View return policy
2100-31-4
C10H12O3
180.203
MFCD01075687
OXOWWPXTTOCKKU-UHFFFAOYSA-N
2-n-propoxybenzoic acid, o-propoxybenzoic acid, benzoic acid, 2-propoxy, 2-propoxy benzoic acid, salicylic acid propyl ether, akos bbb/270, rarechem al bo 0876, propyloxybenzoic acid, 2-propoxybenzoic acid, o-propoxy benzoic acid
539215
2-propoxybenzoic acid
CCCOC1=CC=CC=C1C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 36°C to 40°C
Quantity 1 g
Beilstein 3089998
Formula Weight 180.2
Percent Purity ≥98%
Chemical Name or Material 2-n-Propoxybenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'