missing translation for 'onlineSavingsMsg'
Learn More

2-n-Pentylfuran, 98%

Product Code. 11454693
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454693 10 g 10g
11464693 50 g 50g
11329579 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454693 Supplier Thermo Scientific Alfa Aesar Supplier No. A11607.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3777-69-3 | C9H14O | 138.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3777-69-3
Molecular Formula C9H14O
Molecular Weight (g/mol) 138.21
MDL Number MFCD00036497
InChI Key YVBAUDVGOFCUSG-UHFFFAOYSA-N
Synonym furan, 2-pentyl, 2-n-pentylfuran, 2-amylfuran, furan, pentyl, pentylfuran, natural, unii-6i0qaj1jzq, 2-pentyl furan, fema no. 3317, 6i0qaj1jzq
PubChem CID 19602
IUPAC Name 2-pentylfuran
SMILES CCCCCC1=CC=CO1

Specifications

Density 0.88
Boiling Point 167°C
Flash Point 45°C (113°F)
Refractive Index 1.45
Quantity 10 g
UN Number UN1993
Beilstein 107854
Formula Weight 138.21
Percent Purity 98%
Chemical Name or Material 2-n-Pentylfuran
compliance-icons compliance-icons
Product Identifier
  • 2-(n-Pentyl)furan
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P330-Rinse mouth.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 2-(n-Pentyl)furan

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.