missing translation for 'onlineSavingsMsg'
Learn More

2-n-Octylthiophene, 98%

Product Code. 11480274
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480274 25 g 25g
11470274 5 g 5g
11319569 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480274 Supplier Thermo Scientific Alfa Aesar Supplier No. A13672.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 880-36-4 | C12H20S | 196.352 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 880-36-4
Molecular Formula C12H20S
Molecular Weight (g/mol) 196.352
MDL Number MFCD00041015
InChI Key GIFWAJGKWIDXMY-UHFFFAOYSA-N
Synonym 2-n-octylthiophene, thiophene, 2-octyl, octylthiophene, acmc-209qqw
PubChem CID 70153
IUPAC Name 2-octylthiophene
SMILES CCCCCCCCC1=CC=CS1

Specifications

Density 0.921
Boiling Point 127°C to 129°C (12 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.492
Quantity 25 g
Beilstein 114488
Formula Weight 196.36
Percent Purity 98%
Chemical Name or Material 2-n-Octylthiophene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.