missing translation for 'onlineSavingsMsg'
Learn More

2-(N,N-Dimethylaminomethyl)phenylboronic acid, 98%, Thermo Scientific Chemicals

Product Code. 10251582
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85107-53-5
Molecular Formula C9H14BNO2
Molecular Weight (g/mol) 179.02
MDL Number MFCD01318999
InChI Key VUDCTLOJEPCNRS-UHFFFAOYSA-N
Synonym 2-dimethylamino methyl phenyl boronic acid, 2-n,n-dimethylaminomethyl phenylboronic acid, 2-dimethylamino methyl phenylboronic acid, boronic acid, 2-dimethylamino methyl phenyl, 2-n,n-dimethylaminomethyl benzeneboronic acid, 2-n,n-dimethylaminomethyl phenyl-boronic acid, 2-dimethylamino methyl phenyl boranediol, 2-dimethylamino methyl benzeneboronic acid
PubChem CID 2734345
IUPAC Name [2-[(dimethylamino)methyl]phenyl]boronic acid
SMILES B(C1=CC=CC=C1CN(C)C)(O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Formula Weight 179.02
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-(N, N-Dimethylaminomethyl)phenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-(N,N-Dimethylaminomethyl)phenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'