missing translation for 'onlineSavingsMsg'
Learn More

2-n-Hexyloxybenzaldehyde, 96%, Thermo Scientific Chemicals

Product Code. 11392816
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392816 10 g 10g
11302826 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392816 Supplier Thermo Scientific Alfa Aesar Supplier No. L08988.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7162-59-6 | Molecular Formula: C13H18O2 | Molecular Weight: 206.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7162-59-6
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.29
MDL Number MFCD00014131
InChI Key IFOIDROUJIGQAV-UHFFFAOYSA-N
Synonym o-hexyloxybenzaldehyde, 2-hexyloxy benzaldehyde, 2-n-hexyloxybenzaldehyde, 2-hexyloxybenzaldehyde, hexyloxybenzaldehyde, benzaldehyde, 2-hexyloxy, benzaldehyde, o-n-hexyloxy-,
PubChem CID 138951
IUPAC Name 2-hexoxybenzaldehyde
SMILES CCCCCCOC1=CC=CC=C1C=O

Specifications

Boiling Point 132°C to 134°C (0.5 mmHg)
Refractive Index 1.52
Quantity 10 g
Beilstein 2258613
Sensitivity Air Sensitive
Formula Weight 206.29
Percent Purity 96%
Chemical Name or Material 2-n-Hexyloxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.