missing translation for 'onlineSavingsMsg'
Learn More

2-(n-Hexyloxy)ethanol, 98%

Product Code. 11400497
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400497 1 g 1g
11410497 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11400497 Supplier Thermo Scientific Alfa Aesar Supplier No. B22385.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-25-4 | C8H18O2 | 146.23 g/mol

2-(n-Hexyloxy)ethanol is used as solvents in specialty printing inks and coalescing aids in surface coatings, coupling agent, rust remover, adhesives and surface cleaners.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(n-Hexyloxy)ethanol is used as solvents in specialty printing inks and coalescing aids in surface coatings, coupling agent, rust remover, adhesives and surface cleaners.

Solubility
Soluble in alcohol and ether, water (9.46 g/L ).

Notes
Store under inert gas. Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-25-4
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00045997
InChI Key UPGSWASWQBLSKZ-UHFFFAOYSA-N
Synonym 2-hexyloxy ethanol, ethanol, 2-hexyloxy, 2-hexyloxyethanol, ethylene glycol monohexyl ether, hexyl cellosolve, n-hexyl cellosolve, glycol monohexyl ether, cellosolve, n-hexyl, ethylene glycol n-hexyl ether
PubChem CID 8170
SMILES CCCCCCOCCO

Specifications

Melting Point -42°C
Density 0.888
Boiling Point 208°C
Flash Point 94°C (201°F)
Linear Formula CH3(CH2)5OCH2CH2OH
Refractive Index 1.433
Quantity 1 g
UN Number UN2922
Beilstein 1734691
Solubility Information Soluble in alcohol and ether,water (9.46g/L ).
IUPAC Name 2-hexoxyethanol
Formula Weight 146.23
Percent Purity 99%
Chemical Name or Material 2-(n-Hexyloxy)ethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.