missing translation for 'onlineSavingsMsg'
Learn More

2-n-Hexyldecanoic acid, 97%

Product Code. 11308695
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11308695 25 g 25g
11318695 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11308695 Supplier Thermo Scientific Alfa Aesar Supplier No. L12238.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25354-97-6 | C16H32O2 | 256.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25354-97-6
Molecular Formula C16H32O2
Molecular Weight (g/mol) 256.43
MDL Number MFCD00059651
InChI Key JMOLZNNXZPAGBH-UHFFFAOYSA-N
Synonym decanoic acid, 2-hexyl, hexyldecanoic acid, c16 guerbet fatty acid, 2-hexyldecansaeure, 7-pentadecanecarboxylic acid, 2-hexyldecansaeure german, 2-hexyl-decanoic acid, 2-n-hexyldecanoic acid, 2-hexyldecanoicacid, isocarb 16
PubChem CID 32912
IUPAC Name 2-hexyldecanoic acid
SMILES CCCCCCCCC(CCCCCC)C(=O)O

Specifications

Melting Point 10°C
Density 0.88
Boiling Point 165°C to 168°C (2 mmHg)
Flash Point 174°C (345°F)
Refractive Index 1.446
Quantity 25 g
Beilstein 1777217
Merck Index 14,4709
Formula Weight 256.43
Percent Purity 97%
Chemical Name or Material 2-n-Hexyldecanoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.