missing translation for 'onlineSavingsMsg'
Learn More

2-n-Heptylfuran, 97%

Product Code. 11493284
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11493284

Brand: Thermo Scientific Alfa Aesar A10604.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3777-71-7 | C11H18O | 166.264 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3777-71-7
Molecular Formula C11H18O
Molecular Weight (g/mol) 166.264
MDL Number MFCD00051820
InChI Key BHTUFJXTYNLISA-UHFFFAOYSA-N
Synonym 2-n-heptylfuran, furan, 2-heptyl, unii-7w55a39qxm, ccris 6901, fema no. 3401, 2-heptyl furan, furan, heptyl
PubChem CID 19603
IUPAC Name 2-heptylfuran
SMILES CCCCCCCC1=CC=CO1

Specifications

Density 0.863
Boiling Point 96°C to 98°C (30 mmHg)
Flash Point 75°C (167°F)
Refractive Index 1.4525
Quantity 10 g
Formula Weight 166.27
Percent Purity 98%
Chemical Name or Material 2-n-Heptylfuran

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.