missing translation for 'onlineSavingsMsg'
Learn More

2-(N-Fmoc-4-aminobutyl)-1,3-propanediol, TRC

Product Code. 30392105
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392105 500 mg
30391009 250 mg
30389775 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30392105 Supplier Toronto Research Chemicals Supplier No. TRCF620000500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147190-31-6
Molecular Formula C22 H27 N O4
Molecular Weight (g/mol) 369.45
Synonym (9H-Fluoren-9-yl)methyl (6-hydroxy-5-(hydroxymethyl)hexyl)carbamate
IUPAC Name 9H-fluoren-9-ylmethyl N-[6-hydroxy-5-(hydroxymethyl)hexyl]carbamate
SMILES OCC(CO)CCCCNC(=O)OCC1c2ccccc2c3ccccc13

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C22H27NO4/c24-13-16(14-25)7-5-6-12-23-22(26)27-15-21-19-10-3-1-8-17(19)18-9-2-4-11-20(18)21/h1-4,8-11,16,21,24-25H,5-7,12-15H2,(H,23,26)
Formula Weight 369.194
Chemical Name or Material 2-(N-Fmoc-4-aminobutyl)-1,3-propanediol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.