missing translation for 'onlineSavingsMsg'
Learn More

2-n-Butylthiophene, 98+%

Product Code. 11410703
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410703 5 g 5g
11420703 25 g 25g
11430703 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410703 Supplier Thermo Scientific Alfa Aesar Supplier No. A12740.06
  • ONLINE EXCLUSIVE PRICE
    £31.00
    £27.65 / 5g
    Save £3.35
    10% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1455-20-5 | C8H12S | 140.244 g/mol

2-n-Butylthiophene is an inhibitor of DMH-induced aberrant crypt formation. It is used as a Glutathione S-transferase inducer. It is used as a primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-n-Butylthiophene is an inhibitor of DMH-induced aberrant crypt formation. It is used as a Glutathione S-transferase inducer. It is used as a primary and secondary intermediate.

Solubility
Soluble in acetone and DMSO.

Notes
Storage temperature: 2 - 4°Cand Keep container tightlyclosed in a dryand well-ventilated place. Incompatible to Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1455-20-5
Molecular Formula C8H12S
Molecular Weight (g/mol) 140.244
MDL Number MFCD00022534
InChI Key MNDZHERKKXUTOE-UHFFFAOYSA-N
Synonym 2-n-butylthiophene, thiophene, 2-butyl, 2-n-butyl thiophene, ccris 6900, 2-butyl-thiophene, thiophene, butyl, acmc-20c3ak
PubChem CID 73818
IUPAC Name 2-butylthiophene
SMILES CCCCC1=CC=CS1

Specifications

Density 0.953
Boiling Point 181°C to 182°C
Flash Point 56°C (133°F)
Odor Recognizable
Refractive Index 1.502
Quantity 5 g
UN Number UN1993
Beilstein 107046
Solubility Information Soluble in acetone and DMSO.
Formula Weight 140.25
Percent Purity ≥98%
Chemical Name or Material 2-n-Butylthiophene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.