missing translation for 'onlineSavingsMsg'
Learn More

2-n-Butoxyethanol, 99%

Product Code. 11419116
Change view
Click to view available options
Quantity:
10,000 mL
2500 mL
500 mL
Unit Size:
10000mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419116 10,000 mL 10000mL
11499106 500 mL 500mL
11409116 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419116 Supplier Thermo Scientific Alfa Aesar Supplier No. A17976.0D
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-76-2 | C6H14O2 | 118.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Used in industry for degreasing and detergent applications.2-n-Butoxyethanol is used in the industry for degreasing and detergent applications. It acts as a surfactant in domestic and industrial products. It is an active component of fracturing fluids, drilling stabilizers and oil slick dispersants. It is also used as a solvent for paints, surface coatings, cleaning products and inks. Further, it is employed as a food additive, antimicrobial agent, defoamer, stabilizer and adhesive. In addition to this, it is used as a whiteboard cleaner, liquid soap, cosmetic, dry cleaning solution, lacquer, varnishe and latex paint.

Solubility
Miscible with water, ethyl alcohol, ethyl ether, acetone, n-heptane benzene, mineral oil, aromatic paraffin and halogenated hydrocarbons. Slightly miscible with carbon tetrachloride.

Notes
Excellent Hydrophilic-lipophilic balance (HLB).Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, metals, aluminum and alkalies.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-76-2
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
MDL Number MFCD00002884
InChI Key POAOYUHQDCAZBD-UHFFFAOYSA-N
Synonym butoxyethanol, butyl glycol, butyl cellosolve, ethylene glycol monobutyl ether, n-butoxyethanol, ethanol, 2-butoxy, ethylene glycol butyl ether, butyl oxitol, glycol butyl ether, glycol ether eb
PubChem CID 8133
ChEBI CHEBI:63921
IUPAC Name 2-butoxyethanol
SMILES CCCCOCCO

Specifications

Melting Point -79°C
Density 0.902
Boiling Point 171°C to 173°C
Flash Point 64°C (147°F)
Assay Percent Range 0.98
Odor Mild
Linear Formula CH3(CH2)3OCH2CH2OH
Refractive Index 1.419
Quantity 10,000 mL
UN Number UN2810
Beilstein 1732511
Merck Index 14,1559
Solubility Information Miscible with water,ethyl alcohol,ethyl ether,acetone,n-heptane benzene,mineral oil,aromatic paraffin and halogenated hydrocarbons. Slightly miscible with carbon tetrachloride.
Formula Weight 118.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-n-Butoxyethanol
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.