missing translation for 'onlineSavingsMsg'
Learn More

2-[N-(9-Fluorenylmethoxycarbonyl)-N-decylamino]ethanal, TRC

Product Code. 30395865
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395865 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395865 Supplier Toronto Research Chemicals Supplier No. TRCF47242025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[N-(9-Fluorenylmethoxycarbonyl)-N-decylamino]ethanal
CAS 239088-22-3
Molecular Formula C27H35NO3
Quantity 25 mg
Synonym N-Decyl-N-(2-oxoethyl)-9H-fluoren-9-ylmethyl Ester, Decyl(2-oxoethyl)-carbamic Acid 9H-Fluoren-9-ylmethyl Ester
InChI Formula InChI=1S/C27H35NO3/c1-2-3-4-5-6-7-8-13-18-28(19-20-29)27(30)31-21-26-24-16-11-9-14-22(24)23-15-10-12-17-25(23)26/h9-12,14-17,20,26H,2-8,13,18-19,21H2,1H3
SMILES CCCCCCCCCCN(CC=O)C(=O)OCC1c2ccccc2c3ccccc13
IUPAC Name 9H-fluoren-9-ylmethyl N-decyl-N-(2-oxoethyl)carbamate
Molecular Weight (g/mol) 421.57
Formula Weight 421.2617
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.