missing translation for 'onlineSavingsMsg'
Learn More

2-(Morpholinothio)benzothiazole, TRC

Product Code. 30482577
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30482577 2.5 g 2.5g
30471582 250 mg 250mg
30465194 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30482577 Supplier Toronto Research Chemicals Supplier No. TRCM7327832.5G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-77-2
Molecular Formula C11H12N2OS2
Molecular Weight (g/mol) 252.35
Synonym Benzothiazole, 2-(4-morpholinylthio)- (ACI), 2-(4-Morpholinylthio)benzothiazole (ACI), Benzothiazole, 2-(morpholinothio)- (6CI, 7CI, 8CI), Morpholine, 4-(2-benzothiazolylthio)- (9CI), 2-(4-Morpholinothio)benzothiazole, 2-(4-Morpholinylmercapto)benzothiazole, 2-(Morpholin-4-ylsulfanyl)-1,3-benzothiazole, 2-(Morpholinothio)benzothiazole, 2-Benzothiazolesulfenemorpholide, 2-Benzothiazolesulfenic acid morpholide, 2-Benzothiazolyl N-morpholino sulfide, 2-Benzothiazolylsulfenylmorpholine, 4-(2-Benzothiazolylthio)morpholine, Accel NS, Accelerator NS, Amax, Cure-rite OBTS, Delac MOR, MBS, MBS 80, Meramid M, N,N-(Oxydiethylene)benzothiazole-2-sulfenamide, N-(Oxidiethylene)-2-benzothiazolyl sulfenamide, N-Morpholino-2-benzothiazolylsulfenamide, N-Oxydiethyl-2-benzothiazolesulfenamide, N-Oxydiethylene-2-benzothiazolylsulfenamide, NOBS, NOBS Special, Nocceler MSA, Nocceler MSA-G, NS, NSC 70078, OBTS, Pilcure MOR, Rhenogran MBS 80, SA-M, Sanceler NOB, Santocure MBS, Santocure MOR, Soxinol NBS, Sulfenamide M, Sulfenax MOR, Vulcafor BSM, Vulcafor MBS, Vulkacit MOZ, Vulkacit MOZ/LG
IUPAC Name 4-(1,3-benzothiazol-2-ylsulfanyl)morpholine
SMILES C1CN(CCO1)Sc2nc3ccccc3s2

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C11H12N2OS2/c1-2-4-10-9(3-1)12-11(15-10)16-13-5-7-14-8-6-13/h1-4H,5-8H2
Formula Weight 252.0391
Chemical Name or Material 2-(Morpholinothio)benzothiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.