missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholinopyrimidine-5-carboxylic acid, 95%, Thermo Scientific™

Product Code. 10162834
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10162834 1 g Amber Glass Bottle 1g
10490934 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10162834 Supplier Thermo Scientific Maybridge Supplier No. CC69501DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 253315-05-8
Molecular Formula C9H11N3O3
Molecular Weight (g/mol) 209.205
MDL Number MFCD06589849
InChI Key WMPUFHIOPXMZFI-UHFFFAOYSA-N
Synonym 2-morpholinopyrimidine-5-carboxylic acid, 2-morpholin-4-yl-pyrimidine-5-carboxylic acid, 2-morpholin-4-yl pyrimidine-5-carboxylic acid, 2-4-morpholinyl-5-pyrimidinecarboxylic acid, 5-pyrimidinecarboxylicacid, 2-4-morpholinyl, 5-pyrimidinecarboxylic acid, 2-4-morpholinyl
PubChem CID 3160979
IUPAC Name 2-morpholin-4-ylpyrimidine-5-carboxylic acid
SMILES C1COCCN1C2=NC=C(C=N2)C(=O)O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 209.2
Percent Purity 95%
Chemical Name or Material 2-Morpholinopyrimidine-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.