missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholinonicotinic acid, 97%, Thermo Scientific™

Product Code. 10262352
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10262352 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10262352 Supplier Thermo Scientific Maybridge Supplier No. CC24601DA
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 423768-54-1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C10H12N2O3
MDL Number MFCD03086193
Quantity 1 g
Synonym 2-morpholinonicotinic acid, 2-morpholin-4-yl pyridine-3-carboxylic acid, 2-4-morpholino nicotinic acid, pubchem19585, 2-morpholinonicotinicacid, 2-morpholin-4-ylnicotinic acid, 2-morpholin-4-yl-nicotinic acid, 2-morpholinopyridine-3-carboxylic acid, 3-pyridinecarboxylicacid,2-4-morpholinyl
InChI Key YDUPMJIUJMCXAL-UHFFFAOYSA-N
SMILES C1COCCN1C2=C(C=CC=N2)C(=O)O
IUPAC Name 2-morpholin-4-ylpyridine-3-carboxylic acid
Molecular Weight (g/mol) 208.217
PubChem CID 2776568
Formula Weight 208.22
Percent Purity 97%
Chemical Name or Material 2-Morpholinonicotinic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.