missing translation for 'onlineSavingsMsg'
Learn More

[2-(Morpholinomethyl)phenyl]methanol, 97%, Thermo Scientific™

Product Code. 10072354
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10072354 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10072354 Supplier Thermo Scientific Maybridge Supplier No. CC48609DA
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 91271-63-5
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C12H17NO2
MDL Number MFCD03211251
Quantity 1 g
Synonym 2-morpholinomethyl phenyl methanol, 2-morpholin-4-ylmethyl phenyl methanol, 2-morpholinomethyl-benzylalkohol, o-morpholin-1-yl methyl benzyl alcohol, benzenemethanol, 2-4-morpholinylmethyl, 2-morpholin-4-yl methyl phenyl methanol, 2-morpholin-4-ylmethyl phenyl methan-1-ol, 2-morpholin-4-yl-methyl-phenyl-methanol hydrochloride, 2-morpholin-4-ylmethyl phenyl methanol saltdata: hcl
InChI Key JOBIKMKNOFLXRQ-UHFFFAOYSA-N
SMILES C1COCCN1CC2=CC=CC=C2CO
IUPAC Name [2-(morpholin-4-ylmethyl)phenyl]methanol
Molecular Weight (g/mol) 207.273
PubChem CID 693121
Formula Weight 207.27
Percent Purity 97%
Chemical Name or Material [2-(Morpholinomethyl)phenyl]methanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.