missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholinoisonicotinic acid, 97%, Thermo Scientific™

Product Code. 10163264
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10163264 1 g Amber Glass Bottle 1g
10101913 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10163264 Supplier Thermo Scientific Maybridge Supplier No. CC43101DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 295349-64-3
Molecular Formula C10H12N2O3
Molecular Weight (g/mol) 208.217
MDL Number MFCD03411666
InChI Key OVOIVNZYILYGKU-UHFFFAOYSA-N
Synonym 2-morpholinoisonicotinic acid, 2-morpholin-4-ylisonicotinic acid, 2-morpholin-4-yl-isonicotinic acid, 2-morpholin-4-yl pyridine-4-carboxylic acid, 2-morpholinopyridine-4-carboxylic acid, 2-4-morpholinyl isonicotinic acid, 2-4-morpholinyl-pyridine-4-carboxylic acid, 2-morpholin-4-yl isonicotinic acid, 2-4-morpholinyl-4-pyridinecarboxylic acid, 2-4-morpholinyl-isonitinic acid
PubChem CID 2762479
IUPAC Name 2-morpholin-4-ylpyridine-4-carboxylic acid
SMILES C1COCCN1C2=NC=CC(=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 208.22
Percent Purity 97%
Chemical Name or Material 2-Morpholinoisonicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.