missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholinobenzylamine, 97%, Thermo Scientific™

Product Code. 10036904
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10036904 5 g 5g
10202932 250 mg 250mg
10477422 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10036904 Supplier Thermo Scientific Maybridge Supplier No. CC24313DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 204078-48-8
Molecular Formula C11H16N2O
Molecular Weight (g/mol) 192.26
MDL Number MFCD03086183
InChI Key RNYURNUANACIKS-UHFFFAOYSA-N
Synonym 2-morpholinobenzylamine, 2-morpholinophenyl methanamine, 2-morpholin-4-yl phenyl methanamine, 2-morpholin-4-yl-benzylamine, 2-morpholin-4-yl benzylamine, 2-4-morpholinyl phenyl methanamine, 1-2-morpholin-4-yl phenyl methanamine, 2-morpholin-4-ylphenyl methylamine, 2-morpholino benzylamine, 2-morpholin-4-ylbenzylamine
PubChem CID 2776563
IUPAC Name (2-morpholin-4-ylphenyl)methanamine
SMILES NCC1=CC=CC=C1N1CCOCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 5 g
Formula Weight 192.26
Percent Purity 95%
Chemical Name or Material 2-Morpholinobenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.