missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Morpholinobenzaldehyde, 97%, Maybridge™
£17.08 - £123.00
Chemical Identifiers
CAS | 58028-76-5 |
---|---|
Molecular Formula | C11H13NO2 |
Molecular Weight (g/mol) | 191.23 |
MDL Number | MFCD00662562 |
InChI Key | GTTAEWVBVHSDLX-UHFFFAOYSA-N |
Synonym | 2-morpholinobenzaldehyde, 2-morpholin-4-yl-benzaldehyde, 2-4-morpholino benzaldehyde, 2-morpholin-4-yl benzaldehyde, benzaldehyde, 2-4-morpholinyl, morpholinobenzaldehyde, pubchem16276, acmc-1b05v, 2-morpholinobenzenecarbaldehyde, 4-2-formylphenyl morpholine |
PubChem CID | 2737010 |
IUPAC Name | 2-morpholin-4-ylbenzaldehyde |
SMILES | C1COCCN1C2=CC=CC=C2C=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10019471
|
Maybridge
SEW05383DA |
1g |
£17.08
1g |
Estimated Shipment 03-03-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10321925
|
Maybridge
SEW05383DE |
5g |
£67.31
5g |
Estimated Shipment 03-03-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10374912
|
Maybridge
SEW05383EA |
10g |
£123.00
10g |
Estimated Shipment 03-03-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
58028-76-5 | |
191.23 | |
GTTAEWVBVHSDLX-UHFFFAOYSA-N | |
2737010 | |
C1COCCN1C2=CC=CC=C2C=O |
C11H13NO2 | |
MFCD00662562 | |
2-morpholinobenzaldehyde, 2-morpholin-4-yl-benzaldehyde, 2-4-morpholino benzaldehyde, 2-morpholin-4-yl benzaldehyde, benzaldehyde, 2-4-morpholinyl, morpholinobenzaldehyde, pubchem16276, acmc-1b05v, 2-morpholinobenzenecarbaldehyde, 4-2-formylphenyl morpholine | |
2-morpholin-4-ylbenzaldehyde |
Specifications
2-Morpholinobenzaldehyde | |
1g | |
97.0 | |
C11H13NO2 | |
2-morpholinobenzaldehyde, 2-morpholin-4-yl-benzaldehyde, 2-4-morpholino benzaldehyde, 2-morpholin-4-yl benzaldehyde, benzaldehyde, 2-4-morpholinyl, morpholinobenzaldehyde, pubchem16276, acmc-1b05v, 2-morpholinobenzenecarbaldehyde, 4-2-formylphenyl morpholine | |
C1COCCN1C2=CC=CC=C2C=O | |
191.23 | |
191.23 |
Amber glass bottle | |
58028-76-5 | |
97% | |
MFCD00662562 | |
GTTAEWVBVHSDLX-UHFFFAOYSA-N | |
2-morpholin-4-ylbenzaldehyde | |
2737010 | |
97% |