Learn More
Thermo Scientific Maybridge 2-Morpholinoaniline, 25g, 97%
Brand: Thermo Scientific Maybridge BTB02219EB
2-Morpholinoaniline, 25g, 97%
Code : 31
Additional Details : CAS Number : 5585-33-1 Weight : 0.07500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III