missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholinoaniline, 97%, Thermo Scientific™

Product Code. 10049171
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10049171 10 g 10g
10405692 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10049171 Supplier Thermo Scientific Maybridge Supplier No. BTB02219EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 5585-33-1
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.235
MDL Number MFCD00047408
InChI Key QKWLVAYDAHQMLG-UHFFFAOYSA-N
Synonym 2-morpholinoaniline, 2-morpholin-4-yl aniline, 2-morpholin-4-yl-phenylamine, 2-4-morpholino aniline, 4-2-aminophenyl morpholine, 2-4-morpholinyl aniline, 2-morpholin-4-ylphenylamine, enamine_000782, acmc-1ap2s
PubChem CID 735756
IUPAC Name 2-morpholin-4-ylaniline
SMILES C1COCCN1C2=CC=CC=C2N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 178.23
Percent Purity 97%
Chemical Name or Material 2-Morpholinoaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.