missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2-Morpholino-3-pyridinyl)methanol, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 2-morpholino-3-pyridinyl methanol, 2-morpholin-4-yl pyridin-3-yl methanol, 3-pyridinemethanol,2-4-morpholinyl, 2-morpholinopyridin-3-yl methanol, 3-pyridinemethanol, 2-4-morpholinyl, 2-morpholin-4-ylpyridin-3-yl methanol, 2-morpholin-4-yl-3-pyridyl methan-1-ol |
Formula Weight | 194.23 |
Percent Purity | 97% |
Chemical Name or Material | (2-Morpholino-3-pyridinyl)methanol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'