missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholino-1,3-thiazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10103874
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10103874 250 mg Amber Glass Bottle 250mg
10748055 1 g Amber Glass Bottle 1g
10659505 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10103874 Supplier Thermo Scientific Maybridge Supplier No. CC62401CB
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 332345-29-6
Molecular Formula C8H10N2O3S
Molecular Weight (g/mol) 214.239
InChI Key NILKOMDINYFEEX-UHFFFAOYSA-N
Synonym 2-morpholino-1,3-thiazole-5-carboxylic acid, 2-morpholin-4-yl-1,3-thiazole-5-carboxylic acid, 2-morpholin-4-yl-thiazole-5-carboxylic acid, 5-thiazolecarboxylic acid, 2-4-morpholinyl
PubChem CID 15475835
IUPAC Name 2-morpholin-4-yl-1,3-thiazole-5-carboxylic acid
SMILES C1COCCN1C2=NC=C(S2)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 214.24
Percent Purity 97%
Chemical Name or Material 2-morpholino-1, 3-thiazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.