missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholino-1,3-thiazole-4-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10748655
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10748655 250 mg Amber Glass Bottle 250mg
10566914 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10748655 Supplier Thermo Scientific Maybridge Supplier No. CC62804CB
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 126533-97-9
Molecular Formula C8H10N2O2S
Molecular Weight (g/mol) 198.24
MDL Number MFCD09817537
InChI Key JPJSUXPYPAQDDL-UHFFFAOYSA-N
Synonym 2-morpholinothiazole-4-carbaldehyde, 2-morpholin-4-yl-1,3-thiazole-4-carbaldehyde, 2-morpholino-1,3-thiazole-4-carbaldehyde, 4-thiazolecarboxaldehyde,2-4-morpholinyl, acmc-20ahfg, 4-4-formyl-1,3-thiazol-2-yl morpholine, 2-morpholin-4-yl-1,3-thiazole-4-carboxaldehyde, 2-morpholin-4-yl-1,3-thiazole-4-carboxaldehyd
PubChem CID 15053579
IUPAC Name 2-morpholin-4-yl-1,3-thiazole-4-carbaldehyde
SMILES C1COCCN1C2=NC(=CS2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 198.25
Percent Purity 97%
Chemical Name or Material 2-Morpholino-1, 3-thiazole-4-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.