missing translation for 'onlineSavingsMsg'
Learn More

(2-Morpholino-1,3-thiazol-4-yl)methanol, 97%, Thermo Scientific™

Product Code. 10205724
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10205724 5 g Amber Glass Bottle 5g
10415665 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10205724 Supplier Thermo Scientific Maybridge Supplier No. CC62809DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 126533-96-8
Molecular Formula C8H12N2O2S
Molecular Weight (g/mol) 200.256
MDL Number MFCD09702411
InChI Key QJKKFCRFBAXDQE-UHFFFAOYSA-N
Synonym 2-morpholino-1,3-thiazol-4-yl methanol, 2-morpholin-4-yl-1,3-thiazol-4-yl methanol, 4-thiazolemethanol,2-4-morpholinyl, 2-4-morpholinyl-4-hydroxymethyl thiazole, 2-morpholinothiazol-4-yl methanol, acmc-20doby, 2-morpholinothiazol-4-ylmethanol, 2 4-morpholinyl-4-hydroxymethyl thiazole
PubChem CID 10608093
IUPAC Name (2-morpholin-4-yl-1,3-thiazol-4-yl)methanol
SMILES C1COCCN1C2=NC(=CS2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 200.26
Percent Purity 97%
Chemical Name or Material (2-Morpholino-1, 3-thiazol-4-yl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.