missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TheraPure™ 2'-MOE-G Phosphoramidite, serum vial bottle

Product Code. 30054559 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30054559 1 g 1g
18094100 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30054559 Supplier Thermo Scientific™ Supplier No. 27102202
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

2'-MOE-G CEP is a phosphoramidite containing 5' DMT and the nucleoside guanosine (G) with 2'-O-methoxy-ethyl (MOE) protection, for use in oligonucleotide synthesis.

2'-MOE-G CEP is a phosphoramidite containing 5' DMT and the nucleoside guanosine (G) with 2'-O-methoxy-ethyl (MOE) protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, 2'-O-methoxy-ethyl (MOE) modifications display nuclease resistance, fewer off-target effects, and slightly increased hybridization affinities. These phosphoramidites are commonly used for oligos designed for antisense, siRNA, aptamer, or gRNA-based research. The TheraPure trademark denotes our highest level of phosphoramidite purity, developed with the greatest stringency of high definition analytics and tightly controlled impurity profiles and, therefore, ideal for any therapeutic application. This product is supplied in a 20-mL amber bottle with a serum vial neck style, which is compatible with most Applied Biosystems synthesizers and AKTA synthesizers.

Features:

  • Full chemical name – 5'-O-(4,4'-Dimethoxytrityl)-2'-O-methoxyethyl-N2-isobutyryl-guanosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
  • Molecular formula – C47H60N7O10P
  • Base – G
  • Base Protection – iBu
  • Molecular weight – 914.01 g/mol
  • CAS number – 251647-55-9
  • HPLC purity – ≥99%; M-11 Impurity ≤0.01%
  • 31P NMR purity – ≥99%
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) AKTA Synthesizers, Applied Biosystems Synthesizers
Product Type 2in-MOE-G CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type Guanosine (G)
Reagent Features 2in-MOE modified
Shipping Condition Room Temperature
Purity 99%
Quantity 1 g
Reagent Type Phosphoramidite
Grade TheraPure™
Nucleic Acid Type RNA
Product Line TheraPure
Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.