missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TheraPure™ 2'-MOE-5mC Phosphoramidite, 28-400 finish bottle

Product Code. 17937182 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
2 g
Unit Size:
1g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17937182 1 g 1g
17997112 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17937182 Supplier Thermo Scientific™ Supplier No. 27102011
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

2'-MOE-5mC CEP is a phosphoramidite containing 5' DMT and the nucleoside 5-methyl-cytidine (5mC) with 2'-O-methoxy-ethyl (MOE) protection, for use in oligonucleotide synthesis.

2'-MOE-5mC CEP is a phosphoramidite containing 5' DMT and the nucleoside 5-methyl-cytidine (5mC) with 2'-O-methoxy-ethyl (MOE) protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, 2'-O-methoxy-ethyl (MOE) modifications display nuclease resistance, fewer off-target effects, and slightly increased hybridization affinities. These phosphoramidites are commonly used for oligos designed for antisense, siRNA, aptamer, or gRNA-based research. The TheraPure trademark denotes our highest level of phosphoramidite purity, developed with the greatest stringency of high definition analytics and tightly controlled impurity profiles and, therefore, ideal for any therapeutic application. This product is supplied in a 30-mL amber bottle with a 28-400 finish neck style, which is compatible with most MerMade synthesizers.

Features:
Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-2'-O-methoxyethyl-5-methyl-N4-benzoyl-cytidine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
Molecular formula: C50H60N5O10P
Base: 5mC
Base Protection: Bz
Formula weight: 922.0 g/mol
CAS number: 163759-94-2
HPLC purity: ≥99%; M-11 Impurity ≤0.01%
31P NMR purity: ≥99%

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) MerMade Synthesizers
Product Type 2in-MOE-5mC CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type Cytidine (C)
Reagent Features 2in-MOE modified
Shipping Condition Room Temperature
Purity 99%
Quantity 1 g
Reagent Type Phosphoramidite
Grade TheraPure™
Nucleic Acid Type RNA
Product Line TheraPure
Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.