missing translation for 'onlineSavingsMsg'
Learn More

2-Methylthiophene, 98%

Product Code. 11440693
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440693 500 g 500g
11420693 25 g 25g
11430693 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440693 Supplier Thermo Scientific Alfa Aesar Supplier No. A12736.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 554-14-3 | C5H6S | 98.163 g/mol

2-Methylthiophene, is used as a flavoring agent that naturally occurs in grilled and roasted beef, papaya, cooked shrimp, whiskey, and in trace amounts in roasted chicken;

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylthiophene, is used as a flavoring agent that naturally occurs in grilled and roasted beef, papaya, cooked shrimp, whiskey, and in trace amounts in roasted chicken;

Solubility
Slightly soluble in water. Soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong bases, strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-14-3
Molecular Formula C5H6S
Molecular Weight (g/mol) 98.163
MDL Number MFCD00005451
InChI Key XQQBUAPQHNYYRS-UHFFFAOYSA-N
Synonym thiophene, 2-methyl, 2-methyl thiophene, thiophene, methyl, alpha-methylthiophene, ccris 2936, 2-methyl-thiophene, unii-7115jap77a, 2-methylthiacyclopentadiene, a-thiotolene, pubchem5204
PubChem CID 11126
IUPAC Name 2-methylthiophene
SMILES CC1=CC=CS1

Specifications

Melting Point -63°C
Density 1.017
Boiling Point 112°C to 114°C
Flash Point 7°C (44°F)
Odor Aromatic
Refractive Index 1.52
Quantity 500 g
UN Number UN1993
Beilstein 103734
Solubility Information Slightly soluble in water. Soluble in alcohol.
Formula Weight 98.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Methylthiophene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.