missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)pyridine-3-boronic acid, 95%, Thermo Scientific™

Product Code. p-7029671
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11436300 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11436300 Supplier Thermo Scientific Alfa Aesar Supplier No. H27162.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072944-21-8
Molecular Formula C6H8BNO2S
Molecular Weight (g/mol) 169.005
MDL Number MFCD09037488
InChI Key QLZLLRKAZYHWTE-UHFFFAOYSA-N
Synonym 2-methylthio pyridine-3-boronic acid, 2-methylthio pyridin-3-yl boronic acid, 2-methylsulfanyl pyridin-3-yl boronic acid, 2-methylsulfanyl pyridine-3-boronic acid, 2-methylsulfanyl pyridin-3-ylboronic acid, 2-methylthio-3-pyridinyl boronic acid, acmc-2098of, boronic acid, b-2-methylthio-3-pyridinyl
PubChem CID 44755183
IUPAC Name (2-methylsulfanylpyridin-3-yl)boronic acid
SMILES B(C1=C(N=CC=C1)SC)(O)O

Specifications

Melting Point ∽100°C (decomposition)
Quantity 1 g
Formula Weight 169.01
Percent Purity 95%
Chemical Name or Material 2-(Methylthio)pyridine-3-boronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.