missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)phenol, 97+%

Product Code. 11338245
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338245 25 g 25g
11318245 1 g 1g
11328245 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11338245 Supplier Thermo Scientific Alfa Aesar Supplier No. L10128.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1073-29-6 | C7H8OS | 140.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073-29-6
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.20
MDL Number MFCD00002211
InChI Key SOOARYARZPXNAL-UHFFFAOYSA-N
Synonym 2-methylthio phenol, 2-hydroxythioanisole, 2-methylsulfanyl phenol, 2-hydroxy thioanisole, phenol, 2-methylthio, o-hydroxythioanisole, o-methylthio phenol, phenol, o-methylthio, 2-methylmercaptophenol, 2-methylmercapto phenol
PubChem CID 61261
IUPAC Name 2-methylsulfanylphenol
SMILES CSC1=CC=CC=C1O

Specifications

Density 1.16
Boiling Point 104°C to 106°C (22 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.593
Quantity 25 g
Beilstein 1859745
Formula Weight 140.21
Percent Purity ≥97%
Chemical Name or Material 2-(Methylthio)phenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.