missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)nicotinic acid, 98+%

Product Code. 11362357
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362357 10 g 10g
11352357 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362357 Supplier Thermo Scientific Alfa Aesar Supplier No. L09920.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 74470-23-8 | C7H7NO2S | 169.198 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74470-23-8
Molecular Formula C7H7NO2S
Molecular Weight (g/mol) 169.198
MDL Number MFCD00010101
InChI Key COPSJQVPEUUOKY-UHFFFAOYSA-N
Synonym 2-methylthio nicotinic acid, 2-methylsulfanyl nicotinic acid, 2-methylsulfanyl pyridine-3-carboxylic acid, 2-methylthio pyridine-3-carboxylic acid, 3-pyridinecarboxylic acid, 2-methylthio, 2-methylthio-3-pyridinecarboxylic acid, 2-methylmercapto nicotinic acid, 2-methylthiopyridine-3-carboxylic acid, acmc-20aomm, maybridge1_001445
PubChem CID 262400
IUPAC Name 2-methylsulfanylpyridine-3-carboxylic acid
SMILES CSC1=C(C=CC=N1)C(=O)O

Specifications

Melting Point 215°C to 218°C
Quantity 10 g
Beilstein 4391503
Formula Weight 169.2
Percent Purity ≥98%
Chemical Name or Material 2-(Methylthio)nicotinic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.