missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)ethylamine, 97%, Thermo Scientific Chemicals

Catalog Number 10106713
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
18542-42-2
MFCD00014825
CYWGSFFHHMQKET-UHFFFAOYSA-N
2-methylthio ethylamine, 2-methylthio ethanamine, 2-aminoethyl methyl sulfide, 2-methylsulfanyl ethanamine, ethanamine, 2-methylthio, 2-methylsulfanyl ethan-1-amine, 2-methylthio ethyl amine, ethylamine, 2-methylthio, s-methylcysteamine
87697
2-methylsulfanylethanamine
CSCCN
This item is not returnable. View return policy
18542-42-2
MFCD00014825
CYWGSFFHHMQKET-UHFFFAOYSA-N
2-methylthio ethylamine, 2-methylthio ethanamine, 2-aminoethyl methyl sulfide, 2-methylsulfanyl ethanamine, ethanamine, 2-methylthio, 2-methylsulfanyl ethan-1-amine, 2-methylthio ethyl amine, ethylamine, 2-methylthio, s-methylcysteamine
87697
2-methylsulfanylethanamine
CSCCN

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color White to Brown
Density 0.9740g/mL
Boiling Point 148.0°C
Flash Point 36°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C3H9NS
Linear Formula CH3S(CH2)2NH2
Refractive Index 1.4950 to 1.4970
Quantity 1 g
Specific Gravity 0.974
Molecular Weight (g/mol) 91.18
Formula Weight 91.18
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-(Methylthio)ethylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-(Methylthio)ethylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'