missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)ethanol, 99%

Product Code. 11377445
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377445 5 g 5g
11387445 25 g 25g
11397445 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11377445 Supplier Thermo Scientific Alfa Aesar Supplier No. L05442.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5271-38-5 | C3H8OS | 92.16 g/mol

2-(Methylthio)ethanol is used as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylthio)ethanol is used as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Not miscible in water.

Notes
Do not store together with acids. Store away from Oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5271-38-5
Molecular Formula C3H8OS
Molecular Weight (g/mol) 92.16
MDL Number MFCD00002908
InChI Key WBBPRCNXBQTYLF-UHFFFAOYSA-N
Synonym 2-methylthio ethanol, methylthioethanol, ethanol, 2-methylthio, 2-hydroxyethyl methyl sulfide, 2-methylmercaptoethanol, 2-methylsulfanyl ethanol, 2-methylsulfanyl-ethanol, beta-hydroxyethyl methyl sulfide, 2-methylthioethanol
PubChem CID 78925
ChEBI CHEBI:63861
IUPAC Name 2-methylsulfanylethanol
SMILES CSCCO

Specifications

Density 1.06
Boiling Point 169°C to 171°C
Flash Point 70°C (158°F)
Odor Stench
Linear Formula CH3S(CH2)2OH
Refractive Index 1.4944
Quantity 5 g
UN Number UN3334
Beilstein 1731081
Solubility Information Not miscible in water.
Formula Weight 92.16
Percent Purity 99%
Chemical Name or Material 2-(Methylthio)ethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.