missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)benzoic acid, 98+%, Thermo Scientific Chemicals

Catalog Number 11458553
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
3724-10-5
MFCD00029934
This item is not returnable. View return policy
3724-10-5
MFCD00029934

2-(Methylthio)benzoic acid is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dye stuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylthio)benzoic acid is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dye stuffs.

Solubility
Soluble in Methanol.

Notes
Incompatible with strong oxidizing agents, acids, bases, amines. Store in a cool, dry place. Store in a tightly closed container.
TRUSTED_SUSTAINABILITY
Melting Point 166°C to 168°C
Molecular Formula C8H8O2S
Quantity 25 g
Synonym 2-methylthio benzoic acid, benzoic acid, 2-methylthio, o-methylthio benzoic acid, benzoic acid, o-methylthio, 2-methylsulfanyl benzoic acid, 2-methylmercaptobenzoic acid, 2-carboxyphenyl methyl sulfide, acide methyl-s-2-benzoique, 2-methylmercapto benzoic acid, acide methyl-s-2-benzoique french
Solubility Information Soluble in Methanol.
InChI Key LWJQGKJCZOGGPJ-UHFFFAOYSA-N
SMILES CSC1=CC=CC=C1C(=O)O
IUPAC Name 2-methylsulfanylbenzoic acid
Molecular Weight (g/mol) 168.21
PubChem CID 19498
Formula Weight 168.21
Percent Purity ≥98%
Chemical Name or Material 2- (Methylthio)benzoic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'