missing translation for 'onlineSavingsMsg'
Learn More

2-Methylthiazolidine, 98%

Product Code. 11475097
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475097 5 g 5g
11485097 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11475097 Supplier Thermo Scientific Alfa Aesar Supplier No. B22537.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 24050-16-6 | C4H9NS | 103.183 g/mol

2-Methylthiazolidine is used as sources of low molecular-weight thiol compounds both in EATC and mouse liver homogenate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylthiazolidine is used as sources of low molecular-weight thiol compounds both in EATC and mouse liver homogenate.

Solubility
Slightly soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24050-16-6
Molecular Formula C4H9NS
Molecular Weight (g/mol) 103.183
MDL Number MFCD00053110
InChI Key DQMLFUMEBNHPPB-UHFFFAOYSA-N
PubChem CID 32196
IUPAC Name 2-methyl-1,3-thiazolidine
SMILES CC1NCCS1

Specifications

Boiling Point 76°C to 78°C (36 mmHg)
Flash Point 35°C (95°F)
Quantity 5 g
UN Number UN1993
Synonym 2-methylthiazolidine, thiazolidine, 2-methyl, methyl-2 thiazolidine, methyl-2 thiazolidine french, 2-methyltetrahydrothiazole, acmc-20anpm, thiazolidine,2-methyl, 4-27-00-00661 beilstein handbook reference, +/--2-methylthiazolidine, #
Formula Weight 103.18
Percent Purity 99%
Chemical Name or Material 2-Methylthiazolidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.