missing translation for 'onlineSavingsMsg'
Learn More

2-Methyltetrahydrofuran, ReagentPlus™, ≥99.5%, Honeywell Burdick & Jackson™

Product Code. 15621490 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
4 L
20 L
4 x 4 L
Packaging:
Glass Bottle
Stainless Steel Can
missing translation for 'unitSize'
100mL
2.5 L
20 L
4 L
500mL
Pack of 4
This item is not returnable. View return policy

Product Code. 15621490

missing translation for 'mfr': Honeywell Research Chemicals 15581020L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, contains 150-400 ppm BHT as stabilizer,≥99.5%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-47-9
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD00005367
InChI Key JWUJQDFVADABEY-UHFFFAOYSA-N
Synonym 2-methyltetrahydrofuran, methyltetrahydrofuran, tetrahydrosylvan, tetrahydro-2-methylfuran, furan, tetrahydro-2-methyl, furan, 2-methyl-tetrahydro, 2-methyl tetrahydrofuran, 2-methyl-tetrahydrofuran, 2-methyl-tetrahydro-furan, methyl tetrahydrofuran
PubChem CID 7301
IUPAC Name 2-methyloxolane
SMILES CC1CCCO1

Specifications

Melting Point <-20°C
Color Colorless
Density 0.86g/mL at 25°C
Boiling Point 78°C to 80°C
Quantity 20 L
Flash Point −10°C (14°F)
Packaging Stainless Steel Can
Refractive Index n20/D 1.406(lit.)
UN Number UN2536
Beilstein 102448
Formula Weight 86.13g/mol
Percent Purity ≥99.5%
Grade ReagentPlus
Chemical Name or Material 2-Methyltetrahydrofuran
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methyltetrahydrofuran
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.